Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214195.1 | NP_999360.1 | 1 - 2 | 170 | 100 | |
XM_005663621.3 | XP_005663678.2 | 393 | 100 | |||
XM_005663623.3 | XP_005663680.2 | 479 | 100 | |||
XM_021088708.1 | XP_020944367.1 | 475 | 100 | |||
XM_021088709.1 | XP_020944368.1 | 599 | 100 | |||
XM_021088710.1 | XP_020944369.1 | 292 | 100 | |||
XM_021088711.1 | XP_020944370.1 | 235 | 100 | |||
GenBank mRNA | AF064742.1 | - | 1 - 2 | 170 | 100 | |
AK395394.1 | - | 2 - 3 | 331 | 100 |
Gene Symbol: LOC100153854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005663628.3 | XP_005663685.2 | 276 | 100 | ||
GenBank mRNA | AK230675.1 | - | 2 - 3 | 790 | 100 |
Gene Symbol: LOC100521789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125881.6 | XP_003125929.2 | 521 | 100 | ||
XM_005663625.3 | XP_005663682.2 | 353 | 100 | |||
XM_021090122.1 | XP_020945781.1 | 459 | 100 | |||
XM_021090123.1 | XP_020945782.1 | 509 | 100 | |||
XM_021090124.1 | XP_020945783.1 | 235 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |