Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214202.1 | NP_999367.1 | 1 - 2 | 81 | 88 | |
XM_005664506.3 | XP_005664563.1 | 157 | 88 | |||
GenBank mRNA | AF152021.1 | - | 1 - 2 | 81 | 88 | |
AK233875.1 | - | 1 - 2 | 104 | 88 | ||
AK344803.1 | - | 1 - 2 | 104 | 88 | ||
AK399683.1 | - | 1 - 2 | 135 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |