Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214203.1 | NP_999368.1 | 2 - 3 | 252 | 74 | |
GenBank mRNA | AF148567.1 | - | 2 - 3 | 252 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |