Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214216.1 | NP_999381.1 | 1 - 2 | 59 | 118 | |
XM_021088713.1 | XP_020944372.1 | 174 | 118 | |||
XM_021088714.1 | XP_020944373.1 | 226 | 118 | |||
XM_021088715.1 | XP_020944374.1 | 3204 | 118 | |||
GenBank mRNA | AF056489.1 | - | 1 - 2 | 59 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |