Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214229.1 | NP_999394.1 | 6 - 7 | 1200 | 151 | |
XM_021100475.1 | XP_020956134.1 | 1675 | 151 | |||
XM_021100476.1 | XP_020956135.1 | 1343 | 151 | |||
GenBank mRNA | S80652.1 | - | 6 - 7 | 1200 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Sus scrofa Probe spans exons |