Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214273.1 | NP_999438.1 | 1 - 2 | 535 | 71 | |
GenBank mRNA | JN120792.1 | - | 1 - 2 | 534 | 71 | |
L29342.1 | - | 1 - 2 | 535 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Sus scrofa Probe spans exons |