Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214278.1 | NP_999443.1 | 6 - 7 | 1175 | 106 | |
XM_005667890.3 | XP_005667947.1 | 1469 | 106 | |||
XM_005667891.3 | XP_005667948.1 | 1530 | 106 | |||
XM_005667892.3 | XP_005667949.1 | 1540 | 106 | |||
XM_005667893.3 | XP_005667950.1 | 1369 | 106 | |||
XM_013979999.2 | XP_013835453.1 | 4434 | 106 | |||
XM_021102240.1 | XP_020957899.1 | 1338 | 106 | |||
GenBank mRNA | AF327368.1 | - | 6 - 7 | 1175 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |