Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214279.1 | NP_999444.1 | 5 - 6 | 325 | 92 | |
XM_013978159.2 | XP_013833613.1 | 280 | 92 | |||
GenBank mRNA | AF527991.1 | - | 5 - 6 | 325 | 92 | |
AK232253.1 | - | 5 - 6 | 328 | 92 | ||
AK234031.1 | - | 5 - 6 | 320 | 92 | ||
AK346956.1 | - | 5 - 6 | 328 | 92 | ||
AK348728.1 | - | 4 - 5 | 753 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |