Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214319.1 | NP_999484.1 | 16 - 17 | 1692 | 126 | |
XM_021085862.1 | XP_020941521.1 | 1825 | 126 | |||
GenBank mRNA | AB104654.1 | - | 16 - 17 | 1692 | 126 | |
AK236995.1 | - | 16 - 17 | 1697 | 126 | ||
AK393694.1 | - | 13 - 14 | 1275 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |