Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214350.1 | NP_999515.1 | 8 - 9 | 1273 | 92 | |
XM_021091174.1 | XP_020946833.1 | 2509 | 92 | |||
GenBank mRNA | AB113581.1 | - | 8 - 9 | 1273 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |