Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214353.1 | NP_999518.1 | 2 - 3 | 107 | 124 | |
XM_013985800.2 | XP_013841254.1 | 211 | 191 | |||
GenBank mRNA | AK231482.1 | - | 2 - 3 | 150 | 124 | |
AK344819.1 | - | 2 - 3 | 149 | 124 | ||
AY266299.1 | - | 2 - 3 | 107 | 124 | ||
AY610237.1 | - | 2 - 3 | 125 | 124 | ||
FN401368.1 | - | 2 - 3 | 123 | 124 | ||
JX523418.1 | - | 2 - 3 | 103 | 124 | ||
JX523419.1 | - | 2 - 3 | 103 | 124 | ||
JX523420.1 | - | 2 - 3 | 103 | 124 | ||
JX523421.1 | - | 2 - 3 | 103 | 124 | ||
JX523422.1 | - | 2 - 3 | 103 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |