Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214356.1 | NP_999521.1 | 4 - 5 | 463 | 108 | |
GenBank mRNA | AF139872.1 | - | 1 - 2 | 100 | 108 | |
AJ549925.1 | - | 4 - 5 | 463 | 108 | ||
AK235024.1 | - | 4 - 5 | 462 | 108 | ||
AY609912.1 | - | 4 - 5 | 414 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |