Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214358.1 | NP_999523.1 | 1 - 2 | 113 | 111 | |
GenBank mRNA | AK230590.1 | - | 1 - 2 | 249 | 111 | |
AK240420.1 | - | 1 - 2 | 281 | 111 | ||
AK392196.1 | - | 1 - 2 | 301 | 111 | ||
AK399677.1 | - | 1 - 2 | 251 | 111 | ||
AY610311.1 | - | 1 - 2 | 232 | 111 | ||
U57333.1 | - | 1 - 2 | 113 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |