Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214397.1 | NP_999562.1 | 14 - 15 | 1257 | 67 | |
GenBank mRNA | AB035206.1 | - | 14 - 15 | 1257 | 67 | |
AK233401.1 | - | 5 - 6 | 569 | 67 | ||
AK391785.1 | - | 14 - 15 | 1293 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |