Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214397.1 | NP_999562.1 | 5 - 6 | 301 | 77 | |
GenBank mRNA | AB035206.1 | - | 5 - 6 | 301 | 77 | |
AK391785.1 | - | 5 - 6 | 337 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |