Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214414.1 | NP_999579.1 | 1 - 2 | 217 | 93 | |
XM_021072176.1 | XP_020927835.1 | 195 | 93 | |||
GenBank mRNA | AB052257.1 | - | 1 - 2 | 217 | 93 | |
AK393637.1 | - | 1 - 2 | 195 | 93 | ||
AK393722.1 | - | 1 - 2 | 231 | 93 |
Gene Symbol: LOC100739741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315734.1 | NP_001302663.1 | 1 - 2 | 539 | 93 | |
GenBank mRNA | AK233321.1 | - | 1 - 2 | 397 | 93 | |
AK346315.1 | - | 1 - 2 | 173 | 93 | ||
AK394269.1 | - | 1 - 2 | 197 | 93 | ||
KP687253.1 | - | 1 - 2 | 547 | 93 | ||
KP687256.1 | - | 1 - 2 | 539 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |