Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213873.2 | NP_999038.2 | 8 - 9 | 1028 | 68 | |
GenBank mRNA | EF108301.1 | - | 8 - 9 | 1028 | 68 | |
J04601.1 | - | 8 - 9 | 1045 | 68 | ||
M20920.1 | - | 8 - 9 | 1084 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |