Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130730.1 | NP_001124202.1 | 4 - 5 | 500 | 71 | |
XM_005665854.3 | XP_005665911.1 | 543 | 71 | |||
GenBank mRNA | AB529537.1 | - | 3 - 4 | 477 | 71 | |
AK238335.1 | - | 3 - 4 | 699 | 71 | ||
AK392822.1 | - | 4 - 5 | 494 | 71 | ||
EF530130.1 | - | 4 - 5 | 480 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |