Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130730.1 | NP_001124202.1 | 8 - 9 | 787 | 78 | |
XM_005665854.3 | XP_005665911.1 | 787 | 78 | |||
GenBank mRNA | AK238335.1 | - | 7 - 8 | 986 | 78 | |
AK392822.1 | - | 8 - 9 | 781 | 78 | ||
EF530130.1 | - | 8 - 9 | 767 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |