Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130972.1 | NP_001124444.1 | 2 - 3 | 583 | 125 | |
XM_021089600.1 | XP_020945259.1 | 860 | 125 | |||
GenBank mRNA | EU835903.1 | - | 2 - 3 | 583 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |