Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134346.2 | NP_001127818.1 | 2 - 3 | 472 | 140 | |
GenBank mRNA | EU669078.2 | - | 2 - 3 | 472 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |