Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134354.1 | NP_001127826.1 | 4 - 5 | 441 | 61 | |
XM_005662508.3 | XP_005662565.1 | 628 | 61 | |||
GenBank mRNA | AK343717.1 | - | 4 - 5 | 457 | 61 | |
EU868831.1 | - | 4 - 5 | 441 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |