Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129968.1 | NP_001123440.1 | 2 | 347 | 92 | |
XM_021074558.1 | XP_020930217.1 | 399 | 92 | |||
XM_021074559.1 | XP_020930218.1 | 1009 | 92 | |||
XM_021074560.1 | XP_020930219.1 | 394 | 92 | |||
XM_021074561.1 | XP_020930220.1 | 346 | 92 | |||
XM_021074562.1 | XP_020930221.1 | 694 | 92 | |||
GenBank mRNA | AY585269.2 | - | 2 | 347 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |