Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139472.2 | NP_001132944.1 | 1 | 47 | 81 | |
GenBank mRNA | AK400956.1 | - | 1 | 47 | 81 | |
FJ358606.1 | - | 1 | 50 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |