Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139476.2 | NP_001132948.1 | 1 | 546 | 102 | |
XM_005655570.3 | XP_005655627.1 | 542 | 102 | |||
GenBank mRNA | AK230500.1 | - | 1 | 102 | ||
AK343815.1 | - | 1 | 545 | 102 | ||
AK351723.1 | - | 1 | 561 | 102 | ||
AK389691.1 | - | 1 | 560 | 102 | ||
AK399469.1 | - | 1 | 564 | 102 | ||
FJ358604.1 | - | 1 | 550 | 102 | ||
KU705635.1 | - | 1 | 490 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |