Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142837.1 | NP_001136309.1 | 1 | 454 | 57 | |
GenBank mRNA | AK349085.1 | - | 1 | 484 | 57 | |
FJ455508.1 | - | 1 | 454 | 57 | ||
FJ853390.1 | - | 1 | 454 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |