Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143722.1 | NP_001137194.1 | 9 | 1953 | 68 | |
XM_021066040.1 | XP_020921699.1 | 4058 | 68 | |||
XM_021066041.1 | XP_020921700.1 | 2279 | 68 | |||
XM_021066042.1 | XP_020921701.1 | 4173 | 68 | |||
GenBank mRNA | AK399445.1 | - | 11 | 1490 | 68 | |
FJ458422.1 | - | 9 | 1953 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |