Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144845.1 | NP_001138317.1 | 2 | 548 | 104 | |
XM_005674378.2 | XP_005674435.1 | 721 | 104 | |||
GenBank mRNA | AK346637.1 | - | 4 | 678 | 104 | |
AK346731.1 | - | 4 | 682 | 104 | ||
EU350134.1 | - | 2 | 548 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |