Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145386.1 | NP_001138858.1 | 6 | 2847 | 102 | |
XM_005672408.3 | XP_005672465.1 | 2905 | 102 | |||
GenBank mRNA | EU707806.1 | - | 6 | 2628 | 102 | |
EU707807.1 | - | 6 | 2847 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |