Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134356.1 | NP_001127828.1 | 1 - 2 | 66 | 61 | |
XM_021085573.1 | XP_020941232.1 | 194 | 61 | |||
XM_021085574.1 | XP_020941233.1 | 418 | 61 | |||
GenBank mRNA | AK231683.1 | - | 2 - 3 | 427 | 61 | |
AK234191.1 | - | 2 - 3 | 163 | 61 | ||
AK390890.1 | - | 2 - 3 | 154 | 61 | ||
AK391066.1 | - | 2 - 3 | 169 | 61 | ||
AK396076.1 | - | 2 - 3 | 161 | 61 | ||
EU868834.1 | - | 1 - 2 | 66 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |