Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163411.1 | NP_001156883.1 | 3 - 4 | 341 | 85 | |
XM_021085626.1 | XP_020941285.1 | 775 | 85 | |||
GenBank mRNA | AK348492.1 | - | 3 - 4 | 339 | 85 | |
GQ166958.1 | - | 3 - 4 | 341 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |