Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163411.1 | NP_001156883.1 | 6 - 7 | 954 | 73 | |
GenBank mRNA | AK348492.1 | - | 6 - 7 | 73 | ||
GQ166958.1 | - | 6 - 7 | 954 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |