Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163649.2 | NP_001157121.1 | 4 - 5 | 794 | 66 | |
XM_005656620.3 | XP_005656677.2 | 953 | 66 | |||
XM_005656621.3 | XP_005656678.2 | 1013 | 66 | |||
GenBank mRNA | GQ245762.1 | - | 4 - 5 | 796 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |