Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163997.1 | NP_001157469.1 | 1 - 2 | 34 | 96 | |
XM_005655587.3 | XP_005655644.1 | 133 | 96 | |||
XM_005655589.3 | XP_005655646.1 | 133 | 96 | |||
XM_021091112.1 | XP_020946771.1 | 63 | 116 | |||
GenBank mRNA | AK233447.1 | - | 1 - 2 | 34 | 96 | |
AK238248.1 | - | 1 - 2 | 33 | 96 | ||
AK240054.1 | - | 1 - 2 | 96 | |||
AK240360.1 | - | 1 - 2 | 34 | 96 | ||
AK343746.1 | - | 1 - 2 | 35 | 96 | ||
AK391962.1 | - | 1 - 2 | 41 | 96 | ||
AK392800.1 | - | 1 - 2 | 47 | 96 | ||
AK397509.1 | - | 1 - 2 | 35 | 96 | ||
AK397762.1 | - | 1 - 2 | 48 | 96 | ||
AK399933.1 | - | 1 - 2 | 47 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |