Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164736.1 | NP_001158208.1 | 5 - 6 | 816 | 62 | |
XM_021076535.1 | XP_020932194.1 | 1251 | 62 | |||
GenBank mRNA | GQ421749.1 | - | 5 - 6 | 816 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |