Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167634.1 | NP_001161106.1 | 1 - 2 | 100 | 70 | |
GenBank mRNA | AB529460.1 | - | 1 - 2 | 100 | 70 | |
AK238647.1 | - | 1 - 2 | 306 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |