Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144120.1 | NP_001137592.1 | 5 - 6 | 438 | 65 | |
XM_005664661.3 | XP_005664718.2 | 559 | 65 | |||
GenBank mRNA | EU194251.1 | - | 5 - 6 | 438 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |