Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159613.1 | NP_001153085.1 | 1 - 2 | 260 | 70 | |
GenBank mRNA | AK390517.1 | - | 1 - 2 | 304 | 70 | |
AY610195.1 | - | 1 - 2 | 260 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |