Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159614.1 | NP_001153086.1 | 2 - 3 | 949 | 71 | |
XM_005666124.3 | XP_005666181.1 | 1097 | 71 | |||
GenBank mRNA | KM817029.1 | - | 3 - 4 | 1042 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |