Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159777.1 | NP_001153249.1 | 1 | 654 | 76 | |
GenBank mRNA | FJ875105.1 | - | 1 | 654 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |