Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160081.1 | NP_001153553.1 | 1 - 2 | 361 | 65 | |
GenBank mRNA | FJ875103.1 | - | 1 - 2 | 361 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |