Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162403.1 | NP_001155875.1 | 5 - 6 | 436 | 72 | |
GenBank mRNA | AK232154.1 | - | 5 - 6 | 433 | 72 | |
AK392248.1 | - | 5 - 6 | 448 | 72 | ||
AK398824.1 | - | 5 - 6 | 423 | 72 | ||
AY609773.1 | - | 5 - 6 | 430 | 72 | ||
EU310930.1 | - | 5 - 6 | 436 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |