Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168422.1 | NP_001161894.1 | 5 - 6 | 845 | 83 | |
GenBank mRNA | FN568105.1 | - | 5 - 6 | 845 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |