Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170516.1 | NP_001163987.1 | 2 - 3 | 185 | 73 | |
GenBank mRNA | AK345138.1 | - | 2 - 3 | 318 | 73 | |
AK346812.1 | - | 2 - 3 | 339 | 73 | ||
AK346914.1 | - | 2 - 3 | 314 | 73 | ||
AK347081.1 | - | 2 - 3 | 314 | 73 | ||
AM911345.1 | - | 2 - 3 | 185 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |