Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177920.1 | NP_001171391.1 | 2 - 3 | 217 | 87 | |
GenBank mRNA | GU475012.1 | - | 2 - 3 | 217 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |