Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPATA19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177920.1 | NP_001171391.1 | 4 - 5 | 406 | 73 | |
GenBank mRNA | GU475012.1 | - | 4 - 5 | 406 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |