Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177926.1 | NP_001171397.1 | 3 - 4 | 638 | 77 | |
GenBank mRNA | AK351927.1 | - | 3 - 4 | 830 | 77 | |
AY609832.1 | - | 3 - 4 | 777 | 77 | ||
GQ265874.1 | - | 3 - 4 | 638 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |