Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177926.1 | NP_001171397.1 | 5 - 6 | 1073 | 82 | |
GenBank mRNA | AK239304.1 | - | 6 - 7 | 1218 | 82 | |
AK351927.1 | - | 5 - 6 | 1265 | 82 | ||
AY609832.1 | - | 5 - 6 | 1212 | 82 | ||
GQ265874.1 | - | 5 - 6 | 1073 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |