Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185068.1 | NP_001171997.1 | 4 - 5 | 955 | 77 | |
XM_005655586.3 | XP_005655643.1 | 708 | 77 | |||
XM_021090897.1 | XP_020946556.1 | 944 | 77 | |||
GenBank mRNA | AK240032.1 | - | 4 - 5 | 954 | 77 | |
AK351498.1 | - | 4 - 5 | 935 | 77 | ||
AK394340.1 | - | 2 - 3 | 2393 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |