Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185103.1 | NP_001172032.1 | 8 - 9 | 999 | 66 | |
XM_021079381.1 | XP_020935040.1 | 1060 | 66 | |||
XM_021079382.1 | XP_020935041.1 | 1060 | 66 | |||
XM_021079383.1 | XP_020935042.1 | 1060 | 66 | |||
XM_021079385.1 | XP_020935044.1 | 1077 | 66 | |||
XM_021079386.1 | XP_020935045.1 | 1077 | 66 | |||
GenBank mRNA | AK237711.1 | - | 8 - 9 | 1036 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |